Learn More
Aviva Systems Biology Corp HS6ST3 Antibody - C-terminal region (ARP49081_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49081_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HS6ST3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human HS6ST3. Alternate names: DKFZp761K2315, HS6ST-3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60016