Learn More
Aviva Systems Biology Corp HS747E2A Antibody - middle region (ARP34804_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34804_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HS747E2A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HS747E2A. Alternate names: HS747E2A, bK747E2.1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig
Catalog No. 50-014-52979