Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Human Serum Albumin (HSA), lyophilized powder, Fatty acid free (10 g)
SDP
Catalog No. 502388592
Encompass_Preferred
PAN-Biotech Human Serum Albumin (HSA), Fatty Acid Free lyophilized powder (10 g). APPLICATION: Due to their free hydrophobic region, fatty acid free albumin is used to solubilize lipids in tissue culture. It is also used as a blocking agent in Western blots or ELISA technique and is also an effective cryoprotectant. Fatty acid (FA)-free. FA-free HSA allows for strict control of FAs in cell culture. FA-free HSA maximizes binding sites for the use of specific fatty acids in cell culture. Highly purified product absent of chemical and microbial contamination. Manufactured under conditions certified to ISO 9001. Each donor unit used in the preparation of the product was tested and found negative for Anti-HIV 1/2 and Anti-HCV and nonreactive for HBsAg, HIV-1 Antigen and RPR with kits approved by the FDA. CAS REGISTRY NO.: 70024-90-7. COUNTRY OF ORIGIN: Germany. TYPE: Purified Protein. FATTY ACID CONTENT: Fatty acid free. PROTEASE CONTENT: Not protease free. STORAGE: Store at 2-8°C.
Catalog No. 50-238-8592 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0626110
PAN-Biotech Human Serum Albumin (HSA), Fatty Acid Free lyophilized powder (10 g). APPLICATION: Due to their free hydrophobic region, fatty acid free albumin is used to solubilize lipids in tissue culture. It is also used as a blocking agent in Western blots or ELISA technique and is also an effective cryoprotectant. Fatty acid (FA)-free. FA-free HSA allows for strict control of FAs in cell culture. FA-free HSA maximizes binding sites for the use of specific fatty acids in cell culture. Highly purified product absent of chemical and microbial contamination. Manufactured under conditions certified to ISO 9001. Each donor unit used in the preparation of the product was tested and found negative for Anti-HIV 1/2 and Anti-HCV and nonreactive for HBsAg, HIV-1 Antigen and RPR with kits approved by the FDA. CAS REGISTRY NO.: 70024-90-7. COUNTRY OF ORIGIN: Germany. TYPE: Purified Protein. FATTY ACID CONTENT: Fatty acid free. PROTEASE CONTENT: Not protease free. STORAGE: Store at 2-8°C.