Learn More
Medchemexpress LLC hsa-miR-145-5p mimic 5nmol | 5nmol
SDP
Supplier:  Medchemexpress LLC HYR002825NMOL
Encompass_Preferred
hsa-miR-145-5p mimics are small chemically synthesized double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity
Catalog No. 50-003-21384