Learn More
Medchemexpress LLC hsa-miR-185-3p mimic 5nmol | 5nmol
SDP
Catalog No. 5000321638
Encompass_Preferred
hsa-miR-185-3p mimics are small chemically synthesized double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity
Catalog No. 50-003-21638 Supplier Medchemexpress LLC Supplier No. HYR003455NMOL
hsa-miR-185-3p mimics are small chemically synthesized double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity