Learn More
Medchemexpress LLC hsa-miR-195-3p inhib 5nmol | 5nmol
SDP
Catalog No. 5000322217
Encompass_Preferred
hsa-miR-195-3p inhibitors are chemically-modified oligonucleotides that hybridize with mature miRNAs The miRNA inhibitors have full-length nucleotide 2 -methoxy modification The miRNA inhibitors strongly compete with mature miRNAs to prevent the complementary pairing of miRNAs and their target genes thereby inhibiting miRNAs from functioning
Catalog No. 50-003-22217 Supplier Medchemexpress LLC Supplier No. HYRI003855NMOL
hsa-miR-195-3p inhibitors are chemically-modified oligonucleotides that hybridize with mature miRNAs The miRNA inhibitors have full-length nucleotide 2 -methoxy modification The miRNA inhibitors strongly compete with mature miRNAs to prevent the complementary pairing of miRNAs and their target genes thereby inhibiting miRNAs from functioning