Learn More
Medchemexpress LLC hsa-miR-204-5p inhib 5nmol | 5nmol
SDP
Catalog No. 5000322288
Encompass_Preferred
hsa-miR-204-5p inhibitors are chemically-modified oligonucleotides that hybridize with mature miRNAs The miRNA inhibitors have full-length nucleotide 2 -methoxy modification The miRNA inhibitors strongly compete with mature miRNAs to prevent the complementary pairing of miRNAs and their target genes thereby inhibiting miRNAs from functioning
Catalog No. 50-003-22288 Supplier Medchemexpress LLC Supplier No. HYRI004165NMOL
hsa-miR-204-5p inhibitors are chemically-modified oligonucleotides that hybridize with mature miRNAs The miRNA inhibitors have full-length nucleotide 2 -methoxy modification The miRNA inhibitors strongly compete with mature miRNAs to prevent the complementary pairing of miRNAs and their target genes thereby inhibiting miRNAs from functioning