Learn More
Medchemexpress LLC hsa-miR-296-3p agomi 5nmol | 5nmol
SDP
Catalog No. 5000321516
Encompass_Preferred
hsa-miR-296-3p agomirs are chemically-modified double-strand miRNA mimics with modified mature miRNA strand 2 phosphorothioates at the 5 end 4 phosphorothioates at the 3 end 3 end cholesterol group and full-length nucleotide 2 -methoxy modification They are designed to mimic endogenous miRNAs and recommended for miRNA functional studies Compared with miRNA mimics they exhibits enhanced cellular uptake stability and regulatory activity in vivo
Catalog No. 50-003-21516 Supplier Medchemexpress LLC Supplier No. HYR00505A5NMOL
hsa-miR-296-3p agomirs are chemically-modified double-strand miRNA mimics with modified mature miRNA strand 2 phosphorothioates at the 5 end 4 phosphorothioates at the 3 end 3 end cholesterol group and full-length nucleotide 2 -methoxy modification They are designed to mimic endogenous miRNAs and recommended for miRNA functional studies Compared with miRNA mimics they exhibits enhanced cellular uptake stability and regulatory activity in vivo