Learn More
Medchemexpress LLC hsa-miR-30e-3p mimic 5nmol | 5nmol
SDP
Catalog No. 5000322441
Encompass_Preferred
hsa-miR-30e-3p mimics are small chemically synthesized double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity
Catalog No. 50-003-22441 Supplier Medchemexpress LLC Supplier No. HYR005435NMOL
hsa-miR-30e-3p mimics are small chemically synthesized double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity