Learn More
Medchemexpress LLC hsa-miR-6737-3p mimi 20nmol | 20nmol
SDP
Catalog No. 5000322471
Encompass_Preferred
hsa-miR-6737-3p mimics are small chemically synthesized double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity
Catalog No. 50-003-22471 Supplier Medchemexpress LLC Supplier No. HYR0203120NMOL
hsa-miR-6737-3p mimics are small chemically synthesized double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity