Learn More
Promocell GmbH PromoCell HSAEpC-c 500,000 Human Small Airway Epithelial Cells, cryopreserved
Supplier:  Promocell GmbH C12642
Encompass
Primary Human Small Airway Epithelial Cells (HSAEpC) are isolated from the distal portion of the human respiratory tract in the 1 mm bronchiole area. They stain positive for cytokeratin.The distal respiratory tract mainly consists of pulmonary alveoli, which are spherical outcroppings of the respiratory bronchioles and are the primary sites of gas exchange with the blood. HSAEpC are qualified for functional studies to investigate disorders such as asthma or pulmonary inflammation.Our HSAEpC are now also available from HLA-typed and COPD/Asthma donors.Available formats:* Cryopreserved: Cryogenic vial containing 500.000 viable cells.* Proliferating: >500.000 viable cells shipped in growth medium (T25 flask).
Catalog No. 50-306-048