Learn More
ACROBiosystems Recombinant Protein;Human HSA Protein, low endotoxin;HEK293;1MG;HSA,HSA,ALB,Serum Albumin
Catalog No. 501657673
Encompass_Preferred
Recombinant Protein;Human HSA Protein, low endotoxin;1MG;Human HSA, low endotoxin (HSA-H522a) is expressed from human 293 cells (HEK293). It contains AA Asp 25 - Leu 609 (Accession # NP_000468).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 67.3 kDa. The protein migrates as 66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7673 Supplier ACROBiosystems Supplier No. HSAH522A1MG
Recombinant Protein;Human HSA Protein, low endotoxin;1MG;Human HSA, low endotoxin (HSA-H522a) is expressed from human 293 cells (HEK293). It contains AA Asp 25 - Leu 609 (Accession # NP_000468).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 67.3 kDa. The protein migrates as 66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.