Learn More
Enzo Life Sciences HSC70/HSP70 monoclonal antibody (BB70) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA822F
Encompass_Preferred
Clone: BB70. Host: Mouse. Isotype: IgG2a. Immunogen: Chicken Hsp70/Hsp90 complex. UniProt ID: P08106. Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat, Yeast, Sponge, Rabbit, Housefly, Drosophila. Applications: IHC, IP, WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7524