Learn More
Enzo Life Sciences HSC70/HSP70 monoclonal antibody (BB70) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA822D
Encompass_Preferred
Clone: BB70. Host: Mouse. Isotype: IgG2a. Immunogen: Chicken Hsp70/Hsp90 complex. Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat, Yeast, Sponge, Rabbit, Housefly, Drosophila. Applications: IHC, IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7523