Learn More
Enzo Life Sciences HSC70/HSP70 monoclonal antibody (N27F3-4) (PE conjugate) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA820PEF
Encompass_Preferred
Clone: N27F3-4. Host: Mouse. Isotype: IgG1. Immunogen: Purified Hsc70 (Hsp73) and Hsp70 (Hsp72) isolated from human HeLa cells. Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Plant, Porcine, Monkey, Hamster, Guinea pig, Fish, Dog, Chicken, Bovine, Beluga. Applications: Flow Cytometry. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide.
Catalog No. 50-200-7522