Learn More
Enzo Life Sciences HSC70/HSP70 monoclonal antibody (N27F3-4) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA820D
Encompass_Preferred
Clone: N27F3-4. Host: Mouse. Isotype: IgG1. Immunogen: Purified HSC70 (Hsp73) and HSP70 (Hsp72) isolated from human HeLa cells. UniProt ID: P0DMV8 (HSP70/HSP72), P11142 (HSC70/HSP73). Source: Purified from ascites. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Plant, Porcine, Monkey, Hamster, Guinea pig, Fish, Dog, Chicken, Bovine, Beluga. Applications: Flow Cytometry, IHC (PS), IP, WB. Recommended Dilutions: Flow Cytometry (1:100) Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7520