Learn More
Enzo Life Sciences HSC70/HSP70 monoclonal antibody (N27F3-4) (50µg)
SDP
Supplier:  Enzo Life Sciences ENZABS7160050
Encompass_Preferred
Clone: N27F3-4. Isotype: IgG1κ. Immunogen: Purified HSC70 (Hsp73) and HSP70 (Hsp72) isolated from human HeLa cells. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Plant, Porcine, Monkey, Hamster, Guinea pig, Fish, Dog, Chicken, Bovine, Beluga. Applications: Flow Cytometry, IHC (PS), IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-201-3254