Learn More
Enzo Life Sciences HSC70/HSP70 polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA757F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of rat Hsp70. UniProt ID: P63018. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Yeast, Sheep, Rabbit, Plant, Porcine, Monkey, Hamster, Guinea pig, Fish, Drosophila, Dog, Chicken, Bovine. Applications: IHC (PS), WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7460