Learn More
Enzo Life Sciences HSC70/HSP73 monoclonal antibody (1B5) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA815F
Encompass_Preferred
Clone: 1B5. Host: Rat. Isotype: IgG2a. Immunogen: Native hamster Hsc70 (Hsp73). UniProt: P19378. Source: Purified from ascites. Species Reactivity: Human, Mouse, Rat, Turtle, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine. Applications: ELISA. Flow Cytometry, ICC, IF, IHC (PS), IP, WB, Electron microscopy. Recommended dilutions: Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-582