Learn More
Enzo Life Sciences HSC70/HSP73 (bovine), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPP751D
Encompass_Preferred
Alternative name: Heat shock cognate 71 kDa protein, Heat shock 70 kDa protein 8, HSPA8. Purity: ≥90% (SDS-PAGE; Western blot). MW: ~73kDa. Formulation: Liquid. In 30mM TRIS-HCl, pH 7.5, containing 1.0mM EDTA, 2.0mM DTT, and 300mM sodium chloride. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB Activity assay.
Catalog No. 50-200-7630