Learn More
Enzo Life Sciences HSC70/HSP73 monoclonal antibody (1B5) (PE conjugate) (100µg)
SDP
Supplier:  Enzo Life Sciences ADISPA815PEE
Encompass_Preferred
Clone: 1B5. Host: Rat. Isotype: IgG2a. Immunogen: Native hamster Hsc70 (Hsp73). UniProt ID: P19378. Source: Purified from ascites. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine. Applications: Flow Cytometry. Recommended Dilutions: Flow Cytometry (10µg/ml) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide. Handling: Avoid vigorous centrifugation and vortexing. Do not freeze. Protect from light.
Catalog No. 50-200-7509