Learn More
Enzo Life Sciences HSC70/HSP73 monoclonal antibody (1B5) (biotin conjugate) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA815BD
Encompass_Preferred
Clone: 1B5. Host: Rat. Isotype: IgG2b. Immunogen: Native hamster Hsc70 (Hsp73). Source: Purified from ascites. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine. Applications: ELISA, Flow Cytometry, ICC, IHC, IP, WB Electron microscopy. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide.
Catalog No. 50-200-7507