Learn More
Enzo Life Sciences HSC70/HSP73 polyclonal antibody (1mg)
SDP
Supplier:  Enzo Life Sciences ADISPA816J
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human Hsc70 (Hsp73). The sequence is completely conserved in rat, mouse, hamster and bovine. UniProt ID: P11142. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Bovine. Applications: IHC (PS), WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7512