Learn More
Enzo Life Sciences HSC70/HSP73 polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA819F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human HSC70 (HSP73). The sequence is completely conserved in rat, mouse, bovine, dog, chicken, monkey, and frog. UniProt ID: P11142. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat. Applications: WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7516