Learn More
Enzo Life Sciences HSC70/HSP73 polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA819D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human HSC70 (HSP73). The sequence is completely conserved in rat, mouse, bovine, dog, chicken, monkey, and frog. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat. Applications: WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7515