Learn More
Enzo Life Sciences HSC 70 (plant) monoclonal antibody (1D9) (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA818D
Encompass_Preferred
Clone: 1D9. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant spinach Hsc70 (Hsp73). Source: Purified from mouse ascites. Species Reactivity: Plant, Porcine. Applications: IP, WB. Recommended dilutions: Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-589