Learn More
Enzo Life Sciences HSC70 (plant) monoclonal antibody (5B7) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA817F
Encompass_Preferred
Clone: 5B7. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant spinach Hsc70 (Hsp73). Source: Purified from mouse ascites. Species reactivity: Plant. Applications: IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7514