Learn More
Aviva Systems Biology Corp HSD11B1 Antibody - N-terminal region (ARP45714_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45714_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSD11B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HSD11B1. Alternate names: 11-DH, 11-beta-HSD1, HDL, HSD11, HSD11B, HSD11L, MGC13539, SDR26C1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-59115