Learn More
Aviva Systems Biology Corp HSD11B2 Antibody - N-terminal region (ARP41372_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41372_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSD11B2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HSD11B2. Alternate names: AME, AME1, HSD11K, HSD2, SDR9C3ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-53295