Learn More
Aviva Systems Biology Corp HSD17B11 Antibody - N-terminal region (ARP40843_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40843_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSD17B11. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human HSD17B11. Alternate names: 17-BETA-HSD11, 17-BETA-HSDXI, DHRS8, PAN1B, RETSDR2, SDR16C2, 17BHSD11ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-899