Learn More
Aviva Systems Biology Corp Hsd17b11 Antibody - middle region (ARP40842_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40842_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Hsd17b11. It was validated on Western Blot. . Alternate names: Dhrs8, Pan1b, SDR2, retSDR2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-898