Learn More
Aviva Systems Biology Corp HSD17B14 Antibody - middle region (ARP56865_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56865_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSD17B14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HSD17B14. Alternate names: DHRS10, retSDR3, SDR47C1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62017