Learn More
Aviva Systems Biology Corp HSD17B6 Antibody - N-terminal region (ARP41529_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41529_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HSD17B6. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human HSD17B6. Alternate names: HSE, RODH, SDR9C6ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57894