Learn More
Aviva Systems Biology Corp HSD17B7 Antibody - middle region (ARP61842_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61842_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSD17B7. It was validated on Western Blot. . Alternate names: MGC12523, MGC75018, PRAP, SDR37C1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-483