Learn More
Aviva Systems Biology Corp HSD3B1 Antibody - N-terminal region (ARP41821_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41821_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSD3B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HSD3B1. Alternate names: HSD3B, HSDB3, I, HSDB3A, SDR11E1, 3BETAHSDApplicationWB. Species reactivity: Cow, Goat, Horse, Human, Pig, Rat, Sheep
Catalog No. 50-014-58007