Learn More
Aviva Systems Biology Corp HSD3B2 Antibody - N-terminal region (ARP44239_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44239_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSD3B2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HSD3B2. Alternate names: HSDB, HSDB3, HSD3B, SDR11E2ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rat, Sheep
Catalog No. 50-014-58755