Learn More
Aviva Systems Biology Corp HSD3B7 Antibody - middle region (ARP49934_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49934_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSD3B7. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human HSD3B7. Alternate names: PFIC4, CBAS1, SDR11E3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-465