Learn More
Electron Microscopy Sciences HSE Test Slide S84
SDP
Catalog No. 5026419
Encompass_Preferred
This test slide for calibration in asbestos analysis is made in the UK and is under license from the National Physical Laboratory. It is an epoxy replica of a master slide produced and certified by that laboratory. The replicas are mounted on microscope slides of 1.2 mm thickness with cover glass of 0.17 mm thickness. This information is transferred to the accompanying table, which indicates the maximum phase change passing through test objects to test slide. A satisfactory system will detect block 5. Full details are supplied with the slide.
Catalog No. 50-264-19 Supplier Electron Microscopy Sciences Supplier No. 6803884H
This test slide for calibration in asbestos analysis is made in the UK and is under license from the National Physical Laboratory. It is an epoxy replica of a master slide produced and certified by that laboratory. The replicas are mounted on microscope slides of 1.2 mm thickness with cover glass of 0.17 mm thickness. This information is transferred to the accompanying table, which indicates the maximum phase change passing through test objects to test slide. A satisfactory system will detect block 5. Full details are supplied with the slide.