Learn More
Enzo Life Sciences HSF-1 polyclonal antibody (1mg)
SDP
Supplier:  Enzo Life Sciences ADISPA901J
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human HSF1. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Rabbit, Monkey, Gerbil. Applications: ELISA, IP, WB EMSA. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7585