Learn More
Aviva Systems Biology Corp HSF1 Antibody - C-terminal region (P100769_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100769_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human HSF1. Alternate names: HSTF1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62704