Learn More
Enzo Life Sciences HSF 1 polyclonal antibody (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA901D
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human HSF1. UniProt: Q00613. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Rabbit, Monkey, Gerbil. Applications: ELISA, IP, WB, EMSA. Recommended dilutions: Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-682