Learn More
Enzo Life Sciences HSF1 (human), (recombinant) (His-tag) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPP900F
Encompass_Preferred
Alternative name: HSTF1, Heat shock factor protein 1. Purity: ≥90% (~60% full-length). MW: ~60kDa (predicted), ~70kDa (observed due to His-tag). Formulation: Liquid. In 1x TRIS buffered saline containing 5% glycerol. Source: Produced in E. coli. Human HSF1 is fused at the N-terminus to a His-tag. Long Term Storage: -80°C. Applications: WB in vitro Assay.
Catalog No. 50-200-7655