Learn More
Enzo Life Sciences HSF 1 monoclonal antibody (10H8) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA950F
Encompass_Preferred
Clone: 10H8. Host: Rat. Isotype: IgG1. Immunogen: Recombinant mouse HSF1. UniProt: P38532. Source: Purified from ascites. Species Reactivity: Human, Mouse, Rat, Rabbit, Monkey. Applications: IHC (PS), WB. Recommended dilutions: Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-686