Learn More
Enzo Life Sciences HSF1 (phospho) (human), (recombinant) (His-tag) (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPP902D
Encompass_Preferred
Alternative name: HSTF1, Heat shock factor protein 1. MW: ~60kDa (predicted), ~70kDa (observed due to His-tag and hyperphosphorylation). Purity: ≥90% (~60% full-length with trace upstream kinase). Formulation: Liquid. In 1x TRIS buffered saline containing 5% glycerol. Source: Produced in E. coli. Human phosphorylated HSF1 is fused at the N-terminus to a His-tag. UniProt: Q00613. Application: WB. Recommended dilutions: Western Blot (10ng, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Long Term Storage: -80°C
Catalog No. 50-950-085