Learn More
Medchemexpress LLC HY-103000 10mg Medchemexpress, HSF1A CAS:1196723-93-9 Purity:>98%
SDP
Catalog No. 502028720
Encompass_Preferred
Medchemexpress, HY-103000 10mg HSF1A CAS:1196723-93-9 HSF1A is a cell-permeable activator of heat shock transcription factor 1 (HSF1). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8720 Supplier Medchemexpress LLC Supplier No. HY10300010MG
Medchemexpress, HY-103000 10mg HSF1A CAS:1196723-93-9 HSF1A is a cell-permeable activator of heat shock transcription factor 1 (HSF1). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.