Learn More
TARGETMOL CHEMICALS INC HSF1A 5MG
SDP
Catalog No. 502579946
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HSF1A is a cell-permeable activator of HSF1 that protects mammalian cells against stress-induced apoptosis. purity: 98%
Catalog No. 50-257-9946 Supplier TARGETMOL CHEMICALS INC Supplier No. T41255MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HSF1A is a cell-permeable activator of HSF1 that protects mammalian cells against stress-induced apoptosis. purity: 98%