Learn More
Enzo Life Sciences HSF 2 monoclonal antibody (3E2) (100 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA960E
Encompass_Preferred
Clone: 3E2. Host: Rat. Isotype: IgG. Immunogen: Recombinant mouse HSF2. UniProt: P38533. Source: Purified from mouse ascites. Species Reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Bovine. Applications: WB. Recommended dilutions: Western Blot (4µg/ml, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-688