Learn More
Aviva Systems Biology Corp Hsf2 Antibody - C-terminal region (ARP36897_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36897_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Hsf2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AI661205ApplicationWB. Species reactivity: Human, Mouse, Rabbit, Rat
Catalog No. 50-284-537