Learn More
Aviva Systems Biology Corp HSF2 Antibody - middle region (P100979_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100979_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HSF2. Alternate names: MGC117376, MGC156196, MGC75048, HSF 2, HSTF 2ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62813