Learn More
Aviva Systems Biology Corp HSF2BP Antibody - N-terminal region (ARP34837_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34837_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HSF2BP. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human HSF2BP. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-55581